missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat ARC Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems AF636
This item is not returnable.
View return policy
Description
ARC/NOL3 Polyclonal specifically detects ARC/NOL3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| ARC/NOL3 | |
| Polyclonal | |
| Unconjugated | |
| Apoptosis repressor with CARD, ARCFLJ35304, CARD2, Muscle-enriched cytoplasmic protein, MYC, MYP, NOL3, NOP30, NOPnucleolar protein 3, nucleolar protein 3 (apoptosis repressor with CARD domain), Nucleolar protein of 30 kDa | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 8996 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| NOL3 | |
| KLH-coupled human ARC synthetic peptide CEPEAEAEPEPELE PEPDPEP | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat ARC in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction