missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat BTF3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5747-SP
This item is not returnable.
View return policy
Description
BTF3 Polyclonal specifically detects BTF3 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| BTF3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| basic transcription factor 3, BETA-NAC, BTF3a, BTF3b, NACB, nascent-polypeptide-associated complex beta polypeptide, RNA polymerase B transcription factor 3, transcription factor BTF3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 689 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P20290 | |
| BTF3 | |
| E. coli-derived recombinant human BTF3 Lys46-Asn206 Accession # P20290 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat BTF3 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction