missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Calreticulin Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB38981
This item is not returnable.
View return policy
Description
Calreticulin Monoclonal specifically detects Calreticulin in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Intracellular Staining by Flow Cytometry, Knockout Validated.
Specifications
| Calreticulin | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells | |
| P27797 | |
| CALR | |
| E. coli-derived recombinant human Calreticulin Glu18-Leu417 Accession # P27797 | |
| 100 μg | |
| Primary | |
| Detects human Calreticulin in ELISA. Detects human, mouse and rat Calreticulin in Western blots. | |
| Human | |
| Purified |
| Western Blot, Immunohistochemistry, Flow Cytometry | |
| 681233 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| autoantigen Ro, CALR, Calregulin, calreticulin, cC1qR, CRP55, CRT, Endoplasmic reticulum resident protein 60, epididymis secretory sperm binding protein Li 99n, ERp60, FLJ26680, grp60, HACBP, HEL-S-99n, RO, Sicca syndrome antigen A (autoantigen Ro; calreticulin), SSA, Vasostatin | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 811 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction