missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Cathepsin X/Z/P Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 6 publications
Brand: R&D Systems AF934-SP
This item is not returnable.
View return policy
Description
Cathepsin X/Z/P Polyclonal specifically detects Cathepsin X/Z/P in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunoprecipitation.
Specifications
| Cathepsin X/Z/P | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cathepsin P, Cathepsin Z, CTSX, CTSZ | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 1522 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 3-15 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q9UBR2 | |
| CTSZ | |
| Mouse myeloma cell line NS0-derived recombinant human Cathepsin X/Z/P Ala21-Val303 (Gly23Val, Ser48Thr) Accession # Q9UBR2 | |
| 25 μg | |
| Primary | |
| Detects human Cathepsin X/Z/P in direct ELISAs and Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction