missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat DYRK2 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF5408
This item is not returnable.
View return policy
Description
DYRK2 Polyclonal specifically detects DYRK2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| DYRK2 | |
| Polyclonal | |
| Unconjugated | |
| Q92630 | |
| DYRK2 | |
| E. coli-derived recombinant human DYRK2 Met74-Glu180 Accession # Q92630 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat DYRK2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| dual specificity tyrosine-phosphorylation-regulated kinase 2, dual-specificity tyrosine-(Y)-phosphorylation regulated kinase 2, EC 2.7.12, EC 2.7.12.1, FLJ21217, FLJ21365 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8445 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction