missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat EEA1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF8047-SP
This item is not returnable.
View return policy
Description
EEA1 Polyclonal specifically detects EEA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| EEA1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| early endosome antigen 1, early endosome antigen 1, 162kD, early endosome-associated protein, Endosome-associated protein p162, MST105, MSTP105, ZFYVE2, ZFYVE2MST105, Zinc finger FYVE domain-containing protein 2 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 8411 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q15075 | |
| EEA1 | |
| E. coli-derived recombinant human EEA1 Asn1249-Gln1356 Accession # Q15075 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat EEA1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction