missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Erythropoietin/EPO Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF959
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Polyclonal specifically detects Erythropoietin/EPO in Human, Mouse, Rat samples. It is validated for Western Blot, Neutralization.
Specifications
| Erythropoietin/EPO | |
| Polyclonal | |
| Unconjugated | |
| P07321 | |
| EPO | |
| Chinese hamster ovary cell line CHO-derived recombinant mouse Erythropoietin/EPO, Ala27-Arg192, Accession # P07321 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Erythropoietin/EPO in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant mouse Thymopoietin (Tpo) is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Neutralization | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Neutralization 0.25-1.25 ug/mL | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2056 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction