missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat FKBP38 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3580
This item is not returnable.
View return policy
Description
FKBP38 Polyclonal specifically detects FKBP38 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry.
Specifications
| FKBP38 | |
| Polyclonal | |
| Unconjugated | |
| AAH03739 | |
| FKBP8 | |
| E. coli derived recombinant mouse FKBP38 Met1-Gly326 Accession # AAH03739 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat FKBP38 in Western blots. In Western blots, less than 1% cross-reactivity with recombinant human FKBP-12, -12.6, -13, -25, -51, or -52 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| 38 kDa FK506-binding protein, EC 5.2.1.8,38 kDa FKBP, FK506 binding protein 8, 38kDa, FK506-binding protein 8, FK506-binding protein 8 (38kD), FKBP-38, FKBP38PPIase FKBP8, FKBP8, FKBP-8, FKBPr38, hFKBP38, peptidyl-prolyl cis-trans isomerase FKBP8, rotamase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 23770 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction