missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat FKBP51 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 3 publications
Brand: R&D Systems AF4094
This item is not returnable.
View return policy
Description
FKBP51/FKBP5 Polyclonal specifically detects FKBP51/FKBP5 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.
Specifications
| FKBP51/FKBP5 | |
| Polyclonal | |
| Unconjugated | |
| Q13451 | |
| FKBP5 | |
| E. coli-derived recombinant human FKBP51 Thr2-Val457 Accession # Q13451 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat FKBP51. In Western blots, less than 1% cross-reactivity with recombinant human FKBP12, FKBP13, FKBP38, and FKBP52 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.3 ug/mL, Simple Western 3 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| 51 kDa FKBP, 54 kDa progesterone receptor-associated immunophilin, AIG6,51 kDa FK506-binding protein, Androgen-regulated protein 6, Dit1, FF1 antigen, FK506 binding protein 5, FK506-binding protein 5, FKBP5, FKBP-5, FKBP-51, FKBP51PPIase FKBP5, FKBP54, FKBP54EC 5.2.1.8, HSP90-binding immunophilin, MGC111006, p54, P54,51 kDa FK506-binding protein 5, peptidylprolyl cis-trans isomerase, peptidyl-prolyl cis-trans isomerase FKBP5, PPIase, Ptg-10, Rotamase, T-cell FK506-binding protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2289 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction