missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Glutathione Peroxidase 1/GPX1 Antibody, R&D Systems™

Goat Polyclonal Antibody has been used in 15 publications
Brand: R&D Systems AF3798
This item is not returnable.
View return policy
Description
Glutathione Peroxidase 1/GPX1 Polyclonal specifically detects Glutathione Peroxidase 1/GPX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Knockout Validated.
Specifications
| Glutathione Peroxidase 1/GPX1 | |
| Polyclonal | |
| Unconjugated | |
| P07203 | |
| GPX1 | |
| E. coli-derived recombinant human Glutathione Peroxidase 1 Gly48-Ala201 Accession # P07203 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat GPX1. In Western blots, this antibody shows no cross-reactivity with recombinant human GPX2 or GPX3. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, KnockDown | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunohistochemistry 5-15 ug/mL, Knockout Validated | |
| Cellular glutathione peroxidase, CGPx, EC 1.11.1, EC 1.11.1.9, glutathione peroxidase 1, GPx-1, GPX1, GSHPx-1, GSHPX1, MGC14399, MGC88245 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 2876 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction