Learn More
Human/Mouse/Rat HSP20/HSPB6 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB4200
Description
This antibody was produced using a hybridoma elicited from a mouse immunized with purified, E. coli-derived full length recombinant human Heat Shock Protein 20 kDa (rhHSP20; amino acid 1 - 160; Accession # O14558). The IgG fraction of the tissue culture supernatant was purified by protein G chromatography.
HSP20/HSPB6 Monoclonal specifically detects HSP20/HSPB6 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HSP20/HSPB6 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| O14558 | |
| HSPB6 | |
| E. coli-derived recombinant human HSP20/HSPB6 Met1-Lys160 Accession # O14558 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat HSP20/HSPB6 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 494310 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS, NaCl and Tween™ 20 with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ32389, Heat shock 20 kDa-like protein p20, heat shock protein beta-6, heat shock protein, alpha-crystallin-related, B6, Hsp20, HSPB6 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 126393 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.