missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HSP60 Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems AF1800-SP
This item is not returnable.
View return policy
Description
HSP60 Polyclonal specifically detects HSP60 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western.
Specifications
| HSP60 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 60 kDa chaperonin, Chaperonin 60, CPN60, GROEL, heat shock 60kD protein 1 (chaperonin), heat shock 60kDa protein 1 (chaperonin), Heat shock protein 60, heat shock protein 65, HLD4, Hsp60, HSP-60, HSP60SPG13, HSP65, HSPD1, HuCHA60, Mitochondrial matrix protein P1,60 kDa heat shock protein, mitochondrial, P60 lymphocyte protein, short heat shock protein 60 Hsp60s1, spastic paraplegia 13 (autosomal dominant), SPG13 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 3329 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Simple Western 0.5 - 1 ug/mL | |
| P10809 | |
| HSPD1 | |
| E. coli-derived recombinant human HSP60 Met1-Phe573 Accession # P10809 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat HSP60 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
For Research Use Only
Spot an opportunity for improvement?