missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat HSPA2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6010-SP
This item is not returnable.
View return policy
Description
HSPA2 Monoclonal specifically detects HSPA2 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| HSPA2 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P54652 | |
| HSPA2 | |
| E. coli-derived recombinant human HSPA2 Lys529-Gly616 Accession # P54652 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat HSPA2 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 520608 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Heat shock 70 kDa protein 2, heat shock 70kD protein 2, heat shock 70kDa protein 2, heat shock-related 70 kDa protein 2, HSP70-2, HSP70-3 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3306 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction