missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat IRS1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3978-SP
This item is not returnable.
View return policy
Description
IRS1 Polyclonal specifically detects IRS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| IRS1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| HIRS-1, insulin receptor substrate 1, IRS-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3667 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P35568 | |
| IRS1 | |
| E. coli-derived recombinant human IRS1 Ala801-Ala1020 Accession # P35568 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat IRS1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction