Learn More
Human/Mouse/Rat MDM2/HDM2 Antibody, R&D Systems™
GREENER_CHOICE
Rabbit Polyclonal Antibody has been used in 8 publications
Brand:  R&D Systems AF1244-SP
Product Code. 18737337
  • € 242.00 / 25µg
Description
MDM2/HDM2 Polyclonal specifically detects MDM2/HDM2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MDM2/HDM2
Polyclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ACTFS, HDM2, HDMX, MDM2 oncogene, E3 ubiquitin protein ligase
Rabbit
Affinity Purified
RUO
4193
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Western Blot, Immunohistochemistry
LYOPH
Western Blot 1 ug/mL, Immunohistochemistry 3-15 ug/mL
Q00987
MDM2
E. coli-derived recombinant human MDM2, Asn3-Pro491, Accession # Q00987
25 μg
Primary
Detects human, mouse, and rat MDM2.
Human, Mouse, Rat