missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat mGluR5/1a Antibody, R&D Systems™
Rabbit Polyclonal Antibody
Brand: R&D Systems PPS079
This item is not returnable.
View return policy
Description
mGluR5/1a Polyclonal specifically detects mGluR5/1a in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| mGluR5/1a | |
| Polyclonal | |
| Western Blot 1:1000 dilution, Immunohistochemistry 1:500 dilution | |
| Peptide from the C-terminus region of rat mGluR5 and mGluR1a | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Rat ∼125 kDa mGluR5 and mGluR1a monomers and ∼250 kDa homodimers | |
| For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction