missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Neurexophilin-3/NXPH-3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Marca: R&D Systems AF5098-SP
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Neurexophilin-3 Polyclonal specifically detects Neurexophilin-3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Especificaciones
| Neurexophilin-3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| KIAA1159, neurexophilin 3, Neurexophilin3, NPH3, NPH3neurexophilin-3, NXPH3 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 11248 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| AAI66603 | |
| NXPH3 | |
| Mouse myeloma cell line NS0-derived recombinant rat Neurexophilin-3/NXPH-3 Gln23-Gly252 Accession # AAI66603 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Neurexophilin-3/NXPH-3 in direct ELISAs and Western blots. In direct ELISAs, less than 5% cross-reactivity with recombinant human Neurexophilin-1/NXPH1 and recombinant rat Neurexophilin-1/NXPH1 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido