missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat NLRP10/Pynod/NALP10 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB6606
This item is not returnable.
View return policy
Description
NLRP10/Pynod/NALP10 Monoclonal specifically detects NLRP10/Pynod/NALP10 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NLRP10/Pynod/NALP10 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| Q86W26 | |
| NLRP10 | |
| E. coli-derived recombinant human NLRP10/Pynod/NALP10 Met386-Ile655 Accession # Q86W26 | |
| 100 μg | |
| Primary | |
| Detects human NLRP10/Pynod/NALP10 in direct ELISAs and Western blots. In direct ELISAs, no cross-reactivity with recombinant human (rh) NALP1 (aa 1-323), rhNALP1 (aa 1331-1429), rhNALP2 (aa 1-208), rhNALP3 (aa 3-223), rhNALP3 (aa 540-689), rhNALP5 (aa 469-635), rhNOD1 (aa 830-953), or rhNOD2 (aa 230-450) is observed. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 578502 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CLR11.1, NACHT, leucine rich repeat and PYD containing 10, NALP10, NALP10Pynod, NLR family, pyrin domain containing 10, NOD8, NOD8Nucleotide-binding oligomerization domain protein 8, nucleotide-binding oligomerization domain, leucine rich repeat and pyrin domaincontaining 10, PAN5, Pynod, PYNODNACHT, LRR and PYD domains-containing protein 10 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 338322 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction