missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat NRAGE Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF3835
This item is not returnable.
View return policy
Description
NRAGE/MAGED1 Polyclonal specifically detects NRAGE/MAGED1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NRAGE/MAGED1 | |
| Polyclonal | |
| Unconjugated | |
| Q9Y5V3 | |
| MAGED1 | |
| E. coli-derived recombinant human NRAGE Arg73-Asn254 Accession # Q9Y5V3 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat NRAGE in Western blot. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| DLXIN-1, MAGE tumor antigen CCF, MAGED1, melanoma antigen family D, 1, melanoma-associated antigen D1, Neurotrophin receptor-interacting MAGE homolog, NRAGE, NRAGE MAGE-D1 antigen | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9500 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction