missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Nrf1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB5306
This item is not returnable.
View return policy
Description
Nrf1 Monoclonal specifically detects Nrf1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Nrf1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 2 ug/mL | |
| AAH10623 | |
| NRF1 | |
| E. coli-derived recombinant human Nrf1 Met1-Lys742 Accession # AAH10623 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse and rat Nrf1 at 85 kDa in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 492413 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Alpha palindromic-binding protein, Alpha-pal, EWG, HBZ17, LCR-F1, NFE2L1, NRF-1, nuclear respiratory factor 1, TCF11 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 4899 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction