missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat p27/Kip1 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB22561-SP
This item is not returnable.
View return policy
Description
p27/Kip1 Monoclonal specifically detects p27/Kip1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry.
Specifications
| p27/Kip1 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| P46527 | |
| CDKN1B | |
| E. coli-derived recombinant human p27/Kip1 Met1-Thr198 Accession # P46527 | |
| 25 μg | |
| Primary | |
| Detects human p27/Kip1 in direct ELISAs. Specificity was demonstrated by comparing staining on tissue sections of normal versus p27/Kip1 knockout mice. | |
| Human, Mouse, Rat | |
| Purified |
| Immunohistochemistry | |
| 341116 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CDKN1B, CDKN4, cyclin-dependent kinase inhibitor 1B, cyclin-dependent kinase inhibitor 1B (p27, Kip1), Cyclin-dependent kinase inhibitor p27, Kip1, KIP1P27KIP1, MEN1B, MEN4, p27Kip1 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1027 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction