missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Phospho-GRIN2B/NMDAR2B (Y1472) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems PPS014
This item is not returnable.
View return policy
Description
NMDA R, NR2B Subunit Polyclonal specifically detects NMDA R, NR2B Subunit in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| NMDA R, NR2B Subunit (p tyr1472) | |
| Polyclonal | |
| Western Blot 1:1000 dilution | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2902 | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| Unconjugated | |
| GRIN2B | |
| Phosphopeptide corresponding to amino acid residues surrounding the phospho-Y1472 of NMDA R, NR2B subunit | |
| 100 μL | |
| Primary | |
| Human, mouse, rat ∼180 kDa NMDAR NR2B-subunit protein phosphorylated at Y1472 | |
| For long-term storage, < -20degrees C is recommended. Product is stable at < -20degrees C for at least 1 year. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction