missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Phospho-GSK-3 alpha (S21) Antibody, R&D Systems™

Rabbit Polyclonal Antibody
Brand: R&D Systems AF4125
This item is not returnable.
View return policy
Description
GSK-3 alpha Polyclonal specifically detects GSK-3 alpha in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GSK-3 alpha (p ser21) | |
| Polyclonal | |
| Unconjugated | |
| DKFZp686D0638, EC 2.7.11, EC 2.7.11.26, glycogen synthase kinase 3 alpha, glycogen synthase kinase-3 alpha, GSK3 alpha, GSK-3 alpha, GSK3A | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 2931 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| GSK3A | |
| Phosphopeptide containing human, mouse, and rat GSK-3 alpha S21 site | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat GSK-3 alpha when phosphorylated at S21. This antibody does not detect phosphorylated GSK-3 beta . | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction