missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PLC-gamma 1 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3288
This item is not returnable.
View return policy
Description
PLC-gamma 1 Polyclonal specifically detects PLC-gamma 1 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PLC-gamma 1 | |
| Polyclonal | |
| Unconjugated | |
| P19174 | |
| PLCG1 | |
| E. coli-derived recombinant human PLC-gamma 1 Ser1091-Leu1290 Accession # P19174 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat PLC-gamma 1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| EC 3.1.4.11, gamma 1 (formerly subtype 148), monophosphatidylinositol phosphodiesterase, NCKAP3,1-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma-1, phosphatidylinositol phospholipase C, phosphoinositidase C, phosphoinositide phospholipase C, Phosphoinositide phospholipase C-gamma-1, phospholipase C, gamma 1, phospholipase C-148, Phospholipase C-gamma-1, Phospholipase C-II, PLC11-phosphatidylinositol-4,5-bisphosphate phosphodiesterase gamma 1, PLC148, PLC-148, PLCG1, PLCgamma 1, PLC-gamma-1, PLC-II1-phosphatidyl-D-myo-inositol-4,5-bisphosphate, PPLCA, triphosphoinositide phosphodiesterase | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5335 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction