missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PON3 Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB4345
This item is not returnable.
View return policy
Description
PON3 Monoclonal specifically detects PON3 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| PON3 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL | |
| AAH70374 | |
| PON3 | |
| E. coli-derived recombinant human PON3 Ala30-Leu354 Accession # AAH70374 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat PON3 in Western blots. In Western blots, this antibody does not cross-react with recombinant human (rh) PON1 or rhPON2. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 454929 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 3.1.1.2, EC 3.1.1.81, EC 3.1.8.1, paraoxanase-3, paraoxonase 3, serum paraoxonase/lactonase 3 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5446 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction