missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat PSMA1 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7565-SP
This item is not returnable.
View return policy
Description
PSMA1 Polyclonal specifically detects PSMA1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| PSMA1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| PSMA1 | |
| E. coli-derived recombinant human PSMA1 Gln16-His263 Accession # P25786 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat PSMA1 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| P25786 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 5682 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction