Learn More
Human/Mouse/Rat PTP mu/PTPRM Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB4446-SP
Product Code. 18721069
  • € 214.00 / 25µg
Description
PTP mu/PTPRM Monoclonal specifically detects PTP mu/PTPRM in Human, Mouse, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PTP mu/PTPRM
Monoclonal
LYOPH
Western Blot 1 ug/mL
P28827
PTPRM
Mouse myeloma cell line NS0-derived recombinant human PTPμ/PTPRM Glu21-Lys742 Accession # P28827
25 μg
Primary
Detects human, mouse, and rat PTPμ/PTPRM in Western blots. In Western blots, no cross-reactivity with recombinant human PTPRK, PTPRT, PTPRD, PTPRG, PTPRF (LAR), or DEP1 is observed.
Human, Mouse, Rat
Purified
Western Blot
436502
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
EC 3.1.3.48, hR-PTPu, MGC166994, protein tyrosine phosphatase mu, protein tyrosine phosphatase, receptor type, M, protein tyrosine phosphatase, receptor type, mu polypeptide, Protein-tyrosine phosphatase mu, PTPRL1, PTPRL1R-PTP-MU, PTPRM, receptor-type tyrosine-protein phosphatase mu, RPTPM, R-PTP-mu, RPTPU
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
5797
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b