missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Raf-1 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB4540
Denne vare kan ikke returneres.
Se returpolicy
Beskrivelse
Raf-1 Monoclonal specifically detects Raf-1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Tekniske data
| Raf-1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P04049 | |
| RAF1 | |
| E. coli-derived recombinant human Raf-1 Asn189-Thr353 Accession # P04049 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat Raf-1 at 74 kDa in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 563002 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cRaf, c-Raf, c-Raf-1, CRAFraf proto-oncogene serine/threonine protein kinase, EC 2.7.11, EC 2.7.11.1, NS5, Proto-oncogene c-RAF, RAF, RAF proto-oncogene serine/threonine-protein kinase, Raf1, Raf-1Oncogene RAF1, v-Raf-1, v-raf-1 murine leukemia viral oncogene homolog 1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 5894 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel
Ser du en mulighed for forbedring?Del en indholdskorrektion