missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat RIPK1/RIP1 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB3585-SP
This item is not returnable.
View return policy
Description
RIPK1/RIP1 Monoclonal specifically detects RIPK1/RIP1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry, Knockout Validated.
Specifications
| RIPK1/RIP1 | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 8-25 ug/mL, Knockout Validated | |
| Q13546 | |
| RIPK1 | |
| E. coli-derived recombinant human RIPK1/RIP1 Met1-Asn671 Accession # Q13546 | |
| 25 μg | |
| Primary | |
| Detects human, mouse and rat RIPK1/RIP1 in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry, Immunoassay | |
| 334640 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Cell death protein RIP, EC 2.7.11.1, receptor (TNFRSF)-interacting serine-threonine kinase 1, receptor interacting protein, Receptor-interacting protein 1, receptor-interacting serine/threonine-protein kinase 1, RIP, RIP1, RIP-1, Rip1 kinase, RIPFLJ39204, Serine/threonine-protein kinase RIP | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 8737 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction