missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SHIP Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems MAB2317-SP
This item is not returnable.
View return policy
Description
SHIP Monoclonal specifically detects SHIP in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| SHIP | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
| Q9ES52 | |
| INPP5D | |
| E. coli-derived recombinant mouse SHIP Glu874-Thr941 Accession # Q9ES52 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat SHIP in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunocytochemistry | |
| 257812 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| 145kD, EC 3.1.3, hp51CNMGC142142, Inositol polyphosphate-5-phosphatase of 145 kDa, inositol polyphosphate-5-phosphatase, 145kDa, INPP5D, MGC104855, SHIPMGC142140 | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 3635 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction