missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat SHP-2 Antibody, R&D Systems™

Goat Antibody
Brand: R&D Systems AF10121-100
This item is not returnable.
View return policy
Description
SHP-2/PTPN11 specifically detects SHP-2/PTPN11 in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| SHP-2/PTPN11 | |
| Polyclonal | |
| Western Blot 1 μ/mL | |
| Q06124 | |
| Goat | |
| Antigen Affinity-purified | |
| Primary | |
| Detects human, mouse, and rat SHP-2 in Western blots. | |
| Human, Mouse, Rat | |
| Lyophilized |
| Western Blot | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. | |
| BPTP3, EC 3.1.3.48, MGC14433, protein tyrosine phosphatase, non-receptor type 11, protein tyrosine phosphatase-2, Protein-tyrosine phosphatase 1D, Protein-tyrosine phosphatase 2C, PTP1D, PTP-1D, PTP2C, PTP-2C, PTP2CNS1, PTPN11, SHP2, SHP-2, SHP2CFC, SHPTP2, SH-PTP2, SH-PTP2Noonan syndrome 1, SH-PTP3, tyrosine-protein phosphatase non-receptor type 11 | |
| E. coli-derived recombinant human SHP-2, Glu2-Thr435, Accession No. Q06124 | |
| 100 μg | |
| 5781.0 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70°C as supplied. 1 month, 2 to 8°C under sterile conditions after reconstitution. 6 months, -20 to -70°C under sterile conditions after reconstitution. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction