Learn More
Human/Mouse/Rat SOX1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 42 publications
Brand: R&D Systems AF3369-SP
This item is not returnable.
View return policy
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| SOX1 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6656 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| NP_005977 | |
| SOX1 | |
| E. coli-derived recombinant human SOX1 Asn242-Gly379 (Leu276Ile) Accession # NP_005977 | |
| 25 μg | |
| Primary | |
| Detects human SOX1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) SOX2 is observed. | |
| Human, Mouse, Rat | |
| IgG |