Learn More
Human/Mouse/Rat SOX1 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 42 publications
Brand: R&D Systems AF3369
This item is not returnable.
View return policy
Description
SOX1 Polyclonal specifically detects SOX1 in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| SOX1 | |
| Polyclonal | |
| Unconjugated | |
| NP_005977 | |
| SOX1 | |
| E. coli-derived recombinant human SOX1 Asn242-Gly379 (Leu276Ile) Accession # NP_005977 | |
| 100 μg | |
| Primary | |
| Detects human SOX1 in direct ELISAs and Western blots. In direct ELISAs, less than 15% cross-reactivity with recombinant human (rh) SOX2 is observed. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| SRY (sex determining region Y)-box 1, SRY-related HMG-box gene 1, transcription factor SOX-1 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6656 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |