missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Src Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB3389-SP
This item is not returnable.
View return policy
Description
Src Monoclonal specifically detects Src in Human, Mouse, Rat samples. It is validated for Western Blot.
Specifications
| Src | |
| Monoclonal | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| P12931 | |
| SRC | |
| E. coli-derived recombinant human Src Met1-Ala79 Accession # P12931 | |
| 25 μg | |
| Primary | |
| Detects endogenous human, mouse and rat Src in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot | |
| 327537 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ASV, c-Src, EC 2.7.10, EC 2.7.10.2, pp60c-src, Rous sarcoma, RSVgp4, tyrosine kinase pp60c-src, tyrosine-protein kinase SRC-1, v-src avian sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog, v-src sarcoma (Schmidt-Ruppin A-2) viral oncogene homolog (avian) | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 6714 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction