missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Syntaxin 1A Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF7237-SP
This item is not returnable.
View return policy
Description
Syntaxin 1A Polyclonal specifically detects Syntaxin 1A in Human, Mouse, Rat samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
| Syntaxin 1A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brain Syntaxin, HPC-1, Neuron-specific antigen HPC-1, p35-1, STX1A, STX1P35-1, SYN1A, syntaxin 1A (brain), syntaxin-1A | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 6804 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 0.2 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| Q16623 | |
| STX1A | |
| E. coli-derived recombinant human Syntaxin 1A Met1-Leu165 Accession # Q16623 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Syntaxin 1A in Western blots and detects recombinant human Syntaxin 1A in direct ELISAs. In direct ELISAs, approximately 10% cross-reactivity with recombinant human Syntaxin 1B2 is observed. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction