missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Testican 2/SPOCK2 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF2328-SP
This item is not returnable.
View return policy
Description
Testican 2/SPOCK2 Polyclonal specifically detects Testican 2/SPOCK2 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| Testican 2/SPOCK2 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| FLJ97039, KIAA0275, KIAA0275CWCV, and Kazal-like domains proteoglycan 2, sparc/osteonectin, cwcv and kazal-like domains proteoglycan (testican) 2, SPOCK2 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 9806 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Q92563 | |
| SPOCK2 | |
| Mouse myeloma cell line NS0-derived recombinant human Testican 2/SPOCK2 Glu23-Trp424 Accession # Q92563 | |
| 25 μg | |
| Primary | |
| Detects human, mouse, and rat Testican 2/SPOCK2 in direct ELISAs and human Testican 2/SPOCK2 in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction