missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Tyrosine Hydroxylase Antibody, R&D Systems™
Sheep Polyclonal Antibody
Brand: R&D Systems AF7566
This item is not returnable.
View return policy
Description
Tyrosine Hydroxylase Polyclonal specifically detects Tyrosine Hydroxylase in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry.
Specifications
| Tyrosine Hydroxylase | |
| Polyclonal | |
| Unconjugated | |
| P07101 | |
| TH | |
| E. coli-derived recombinant human Tyrosine Hydroxylase Ala278-Tyr401, predicted Accession # P07101 | |
| 100 μg | |
| Primary | |
| Detects human, mouse and rat Tyrosine Hydroxylase in Western blots. | |
| Human, Mouse, Rat | |
| IgG |
| Western Blot, Immunocytochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Simple Western 10 ug/mL, Immunocytochemistry 5-15 ug/mL | |
| DYT14, DYT5b, EC 1.14.16, EC 1.14.16.2, TH, TYH, TYH dystonia 14, Tyrosine 3-hydroxylase, tyrosine 3-monooxygenase, tyrosine hydroxylase | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 7054 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction