missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Vimentin PE-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems IC2105P
This item is not returnable.
View return policy
Description
Vimentin Monoclonal specifically detects Vimentin in Human, Mouse, Rat samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| Vimentin | |
| Monoclonal | |
| R-PE | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| FLJ36605, VIM, vimentin | |
| Rat | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7431 | |
| Human, Mouse, Rat | |
| Purified |
| Flow Cytometry | |
| 280618 | |
| Intracellular Staining by Flow Cytometry 10 uL/10^6 cells | |
| P08670 | |
| VIM | |
| E. coli-derived recombinant human Vimentin Ser2-Glu466 Accession # P08670 | |
| 100 Tests | |
| Primary | |
| Detects human Vimentin in Western blots. Unconjugated antibody detects mouse and rat Vimentin in immunocytochemistry. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction