missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat Yes Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB32051
This item is not returnable.
View return policy
Description
Yes Monoclonal specifically detects Yes in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Yes | |
| Monoclonal | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 8-25 ug/mL | |
| P07947.3 | |
| YES1 | |
| E. coli-derived recombinant human Yes Met1-Gly88 Accession # P07947.3 | |
| 100 μg | |
| Primary | |
| Detects endogenous human, mouse, and rat Yes in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, Immunohistochemistry | |
| 339827 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| cellular yes-1 protein, c-yes, EC 2.7.10, EC 2.7.10.2, HsT441, P61-YES, Proto-oncogene c-Yes, proto-oncogene tyrosine-protein kinase Yes, v-yes-1 Yamaguchi sarcoma viral oncogene homolog 1, Yamaguchi sarcoma oncogene, Yes, YES1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 7525 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction