missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse RCOR1/CoREST Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF6047-SP
This item is not returnable.
View return policy
Description
RCOR1/CoREST Polyclonal specifically detects RCOR1/CoREST in Human, Mouse samples. It is validated for Western Blot.
Specifications
| RCOR1/CoREST | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CoREST, KIAA0071CORESTRCORREST corepressor, Protein CoREST, RCOR1, REST corepressor 1 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 23186 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UKL0 | |
| RCOR1 | |
| E. coli-derived recombinant human RCOR1/CoREST Ala397-Ser486 Accession # Q9UKL0 | |
| 25 μg | |
| Primary | |
| Detects human and mouse RCOR1/CoREST in direct ELISAs and Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction