missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SEZ6L2/BSRP-A Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF4916-SP
Dieser Artikel kann nicht zurückgegeben werden.
Rückgaberichtlinie anzeigen
Beschreibung
SEZ6L2/BSRP-A Polyclonal specifically detects SEZ6L2/BSRP-A in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Spezifikation
| SEZ6L2/BSRP-A | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| BSRPA, BSRP-A, PSK-1, seizure 6-like protein 2, seizure related 6 homolog (mouse)-like 2, UNQ1903/PRO4349 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 26470 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Q4V9Z5 | |
| SEZ6L2 | |
| Mouse myeloma cell line NS0-derived recombinant mouse SEZ6L2/BSRP-A Leu28-Ser837 Accession # Q4V9Z5 | |
| 25 μg | |
| Primary | |
| Detects human and mouse SEZ6L2/BSRP-A in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant mouse (rm) BSRP-B and rmBSRP-C is observed. | |
| Human, Mouse | |
| IgG |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur