missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse SSEA-3 Alexa Fluor™ 350-conjugated Antibody, R&D Systems™

Rat Monoclonal Antibody
Brand: R&D Systems FAB1434U-100UG
This item is not returnable.
View return policy
Description
SSEA-3 Monoclonal specifically detects SSEA-3 in Human, Mouse samples. It is validated for Flow Cytometry.
Specifications
| SSEA-3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| SSEA3, Stage specific embryonic antigen 3, Stage-specific embryonic antigen 3 | |
| Four to eight cell stage mouse embryos | |
| RUO | |
| Recognizes a carbohydrate epitope of SSEA-3 (4, 5). | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| MC-631 | |
| Alexa Fluor 350 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| Rat | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| IgM |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction