missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Sulfatase-2/SULF2 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7087-SP
This item is not returnable.
View return policy
Description
Sulfatase-2/SULF2 Monoclonal specifically detects Sulfatase-2/SULF2 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| Sulfatase-2/SULF2 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| DKFZp313E091, EC 3.1.6, EC 3.1.6.-, EC 3.1.6.14, extracellular sulfatase Sulf-2, FLJ44488, FLJ90554, hSulf-2, KIAA1247HSULF-2, MGC126411, SULF2, SULF-2, sulfatase 2, Sulfatase2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 55959 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot | |
| 2B4 | |
| Western Blot 1 ug/mL | |
| NP_940998 | |
| SULF2 | |
| Recombinant human Sulfatase-2/SULF2, used to immunize a SULF2-deficient mouse Accession # NP_940998 | |
| 25 μg | |
| Primary | |
| Detects human Sulfatase-2/SULF2 in direct ELISAs and Western blots. | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction