missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse Transferrin Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF3987-SP
This item is not returnable.
View return policy
Description
Transferrin Polyclonal specifically detects Transferrin in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Transferrin | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| TF | |
| Mouse plasma-derived Transferrin | |
| 25 μg | |
| Primary | |
| Detects mouse Transferrin in direct ELISAs. Detects human Transferrin in Western blots. | |
| Human, Mouse | |
| IgG |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Beta-1 metal-binding globulin, DKFZp781D0156, EC 3.4.21, PRO1557, PRO2086, serotransferrin, Siderophilin, TF, transferrin | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 7018 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction