missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse WASF3/WAVE3 Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems AF5515-SP
This item is not returnable.
View return policy
Description
WASF3/WAVE3 Polyclonal specifically detects WASF3/WAVE3 in Human, Mouse samples. It is validated for Western Blot.
Specifications
| WASF3/WAVE3 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| Brush-1, KIAA0900WASP family Verprolin-homologous protein 3, Protein WAVE-3, SCAR3, SCAR3WASP family protein member 3, Verprolin homology domain-containing protein 3, WAS protein family, member 3, WAVE3, WAVE3Brush-1, wisk | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 10810 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot | |
| LYOPH | |
| Western Blot 1 ug/mL | |
| Q9UPY6 | |
| WASF3 | |
| E. coli-derived recombinant human WASF3/WAVE3 Met1-Lys180 Accession # Q9UPY6 | |
| 25 μg | |
| Primary | |
| Detects endogenous human/mouse WASF3/WAVE3 in Western blots. | |
| Human, Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction