missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MRGX2 Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB4727
This item is not returnable.
View return policy
Description
MRGX2 Monoclonal specifically detects MRGX2 in Human samples. It is validated for Flow Cytometry, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
| MRGX2 | |
| Monoclonal | |
| LYOPH | |
| Flow Cytometry 0.25 uL/10^6 cells, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
| Q96LB1 | |
| MRGPRX2 | |
| NS0 mouse myeloma cell line transfected with human MRGX2 Met1-Val330 Accession # Q96LB1 | |
| 100 μg | |
| Primary | |
| Detects human MRGX2. Stains human MRGX2 transfectants but not irrelevant transfectants. | |
| Human | |
| Purified |
| Flow Cytometry, CyTOF | |
| 477533 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| MAS-related GPR, member X2, mas-related G-protein coupled receptor member X2, MRGPRX2, MRGX2G protein-coupled receptor MRGX2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 117194 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction