missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MRP1 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems IC19291R-100UG
This item is not returnable.
View return policy
Description
MRP1 Monoclonal specifically detects MRP1 in Human samples. It is validated for Intracellular Staining by Flow Cytometry.
Specifications
| MRP1 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MRP1 in direct ELISAs and Western blots. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
| Flow Cytometry | |
| QCRL | |
| Intracellular Staining by Flow Cytometry 0.25-1 ug/10^6 cells | |
| ABC29, ABCC, ABCC1, ATP-binding cassette sub-family C member 1, ATP-binding cassette, sub-family C (CFTR/MRP), member 1, EC 3.6.3, EC 3.6.3.44, GS-X, Leukotriene C(4) transporter, LTC4 transporter, MRP1DKFZp781G125, MRPDKFZp686N04233, multidrug resistance associated protein 1, multidrug resistance protein, multidrug resistance-associated protein 1, multiple drug resistance protein 1, multiple drug resistance-associated protein | |
| H69AR human small cell lung cancer cell line membrane extracts | |
| RUO | |
| 4363 | |
| Human | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction