missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human MRVI1 (aa 457-556) Control Fragment Recombinant Protein

Product Code. 30205857
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30205857 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30205857 Supplier Invitrogen™ Supplier No. RP89975

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Recombinant Protein

Highest antigen sequence indentity to the following orthologs: Mouse (89%), Rat (89%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-52592 (PA5-52592. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

This gene is similar to a putative mouse tumor suppressor gene (Mrvi1) that is frequently disrupted by mouse AIDS-related virus (MRV). The encoded protein, which is found in the membrane of the endoplasmic reticulum, is similar to Jaw1, a lymphoid-restricted protein whose expression is downregulated during lymphoid differentiation. Studies in mouse suggest that MRV integration at Mrvi1 induces myeloid leukemia by altering the expression of a gene important for myeloid cell growth and/or differentiation, and thus this gene may function as a myeloid leukemia tumor suppressor gene. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene, few of which initiate translation at a non-AUG (CUG) start site.
TRUSTED_SUSTAINABILITY

Specifications

Accession Number Q9Y6F6
Concentration ≥5.0 mg/mL
For Use With (Application) Blocking Assay, Control
Formulation 1 M urea, PBS with no preservative; pH 7.4
Gene ID (Entrez) 10335
Name Human MRVI1 (aa 457-556) Control Fragment
Quantity 100 μL
Regulatory Status RUO
Gene Alias BB115629; inositol 1,4,5-triphosphate receptor associated 1; inositol 1,4,5-triphosphate receptor associated 1; protein MRVI1; inositol 1,4,5-triphosphate receptor-associated cGMP kinase substrate; inositol 1,4,5-trisphosphate receptor-associated cGMP kinase substrate; IP3 receptor associated cGMP kinase substrate; IP3 receptor associated cGMP kinase substrate A; IP3 receptor associated cGMP kinase substrate B; IP3R-associated cGMP kinase substrate; IRAG; IRAG1; JAW1L; JAW1-related protein MRVI1; MRV integration site 1; MRVI1; murine retrovirus integration site 1 homolog; murine retrovirus integration site 1 protein; Protein MRVI1; retroviral integration site 1; Ris1
Common Name MRVI1
Gene Symbol MRVI1
Conjugate Unconjugated
Species Human
Recombinant Recombinant
Protein Tag His-ABP-tag
Sequence NLVGLKLPDLSEAAEQEKGLPSELSPAIEEEESKSGLDVMPNISDVLLRKLRVHRSLPGSAPPLTEKEVENVFVQLSLAFRNDSYTLESRINQAERERNL
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System E. coli
Form Liquid
Purity or Quality Grade >80% by SDS-PAGE and Coomassie blue staining
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.