missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human MS4A3 Alexa Fluor 405-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB6247V-100UG
This item is not returnable.
View return policy
Description
MS4A3 Monoclonal specifically detects MS4A3 in Human samples. It is validated for Flow Cytometry.
Specifications
| MS4A3 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| NP_006129 | |
| MS4A3 | |
| NS0 mouse myeloma cell line transfected with human MS4A3 Accession # NP_006129 | |
| RUO | |
| 932 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 489433 | |
| Alexa Fluor 405 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| CD20 antigen homolog, CD20 antigen-like protein, CD20L, CD20LIgE receptor beta subunit, hematopoietic cell 4 transmembrane protein, Hematopoietic-specific transmembrane protein 4, HTM4, HTM4IgE receptor beta chain, membrane-spanning 4-domains subfamily A member 3, membrane-spanning 4-domains, subfamily A, member 3 (hematopoieticcell-specific) | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human MS4A3 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction