missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Musashi-2 Alexa Fluor 647-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB3255R-100UG
This item is not returnable.
View return policy
Description
Musashi-2 Monoclonal specifically detects Musashi-2 in Human samples. It is validated for Flow Cytometry.
Specifications
| Musashi-2 | |
| Monoclonal | |
| Alexa Fluor 647 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. | |
| FLJ36569, MGC3245, MSI2, MSI2H, musashi homolog 2 (Drosophila), musashi-2, Musashi2, RNA-binding protein Musashi homolog 2 | |
| E. coli-derived recombinant human Musashi-2 Met1-His328 Accession # Q96DH6 | |
| RUO | |
| 124540 | |
| Human | |
| IgG2b |
| Flow Cytometry | |
| 960121 | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q96DH6 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human Musashi-2 in direct ELISAs. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction